missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Manual transfer of reagents into plates during routine experimental setups can be time-consuming and tedious. Daily preparation of such plates can negatively impact overall productivity. To ease the stress and minimize the error rate, TaqMan™ Array Plates are designed to reduce redundant pipetting steps and minimize loading errors, making them an ideal solution for fast and easy experimental setup. TaqMan™ Array Plates contain high-quality TaqMan™ Gene Expression Assays (TaqMan™ probe and PCR primer sets) in a convenient, dried-down, 96- or 384-well plate format. Extensive tests have found no difference in performance between TaqMan™ Assays supplied in solution in single tubes and assays supplied dried in a TaqMan™ Array Plate
- Extensive collections - choose from over 130 predesigned TaqMan™ Gene Signature Arrays and Plates, or customize your own plate, selecting from any of our 1.3 million inventoried assays across 24 species
- Affordable - minimum order of only one gene signature plate or six custom plates
- Convenient - dried TaqMan probe and primer sets supplied in 96-well plates; just add master mix and sample, and then run
- Quick results - Fast format typically provides results in under an hour
Specifications
Specifications
| Content And Storage | Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C. |
| Form | Dried Down |
| Format | 96-well plate |
| Reaction Speed | Standard |
| Internal Probe Modification | MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') |
| Label or Dye | FAM |
| Product Line | TaqMan |
| Purification Method | Standard |
| Quantity | 1 standard plate |
| Research Category | Cell Cycle & Proliferation, Apoptosis, Signal Transduction, Biomarker-Related Pathway(s), Stem Cell Research, Immunology, Oncology |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?