missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation Symmetry C18 Column

Product Code. 10063494
Encompass
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
40 mm
Particle Size:
3.5 μm
5 μm
7 μm
Unit Size:
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
10063494 100 mm 3 mm 3.5 μm Each
13210130 50 mm 2.1 mm 3.5 μm Each
13280130 150 mm 4.6 mm 3.5 μm Each
13210140 150 mm 2.1 mm 3.5 μm Each
13220140 50 mm 4.6 mm 3.5 μm Each
10170370 50 mm 4.6 mm 5 μm Each
13282780 150 mm 3 mm 3.5 μm Each
13135009 100 mm 40 mm 7 μm Each
13220130 100 mm 2.1 mm 3.5 μm Each
13230130 75 mm 4.6 mm 3.5 μm Each
13240130 100 mm 4.6 mm 3.5 μm Each
13230140 50 mm 2.1 mm 5 μm Each
13240170 30 mm 4.6 mm 3.5 μm Each
13210410 100 mm 2.1 mm 5 μm Each
13240410 50 mm 3 mm 3.5 μm Each
13280410 100 mm 4.6 mm 5 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 10063494 Supplier Waters Corporation Supplier No. 186000696

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactivated silica.

  • Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
  • Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods.
  • No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
  • Available in a variety of particle sizes, chemistries and dimensions.

Specifications

Diameter (Metric) 3 mm
Length (Metric) 100 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 3.5 μm
Pore Size 100 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.