missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation SunFire Prep C18 OBD Column

Product Code. 10136054
Encompass
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
19 mm
30 mm
50 mm
Particle Size:
5 μm
10 μm
Unit Size:
Each
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
10136054 100 mm 30 mm 10 μm Each
13252910 50 mm 50 mm 10 μm Each
13223270 150 mm 50 mm 5 μm Each
10176406 250 mm 30 mm 5 μm Each
13233290 250 mm 50 mm 5 μm Each
13243290 100 mm 50 mm 10 μm Each
13270440 50 mm 19 mm 10 μm Each
13280440 150 mm 19 mm 10 μm Each
13290440 250 mm 19 mm 10 μm Each
13200450 150 mm 30 mm 10 μm Each
13210450 250 mm 30 mm 10 μm Each
13220450 150 mm 50 mm 10 μm Each
13230450 250 mm 50 mm 10 μm Each
13262860 150 mm 30 mm 5 μm Each
13212910 50 mm 50 mm 5 μm Each
13232910 100 mm 50 mm 5 μm Each
13283330 250 mm 19 mm 5 μm Each
13253230 50 mm 30 mm 10 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 10136054 Supplier Waters Corporation Supplier No. 186003971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SunFire C18 columns are used for general purpose method development at low pH.

  • The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.

Specifications

Diameter (Metric) 30 mm
Length (Metric) 100 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 10 μm
Pore Size 100 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.