missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUMO-3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10169542
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10169542 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10169542 Supplier Invitrogen™ Supplier No. 401900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

SUMO-3 Polyclonal Antibody for Western Blot

Small ubiquitin-related modifier proteins are members of the ubiquitin-like protein family. SUMO-3 regulates Amyloid beta generation and may be critical in the onset or progression of Alzheimer's disease. SUMO-2/3 forms poly-. SUMO-1, 2, and 3 proteins localize to the nuclear membrane, nuclear bodies, and cytoplasm, respectively. SUMOylation is essential for mammalian cell cycle regulation, SUMO-2/3 localizes to centromeres and condensed chromosomes where binds CENP-E.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO-3
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene SUMO3
Gene Accession No. P55854
Gene Alias 2810014B19Rik; D10Ertd345e; small ubiquitin like modifier 3; small ubiquitin-like modifier 3; small ubiquitin-related modifier 3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); Smt3A; Smt3B; SMT3H1; SUMO-2; Sumo3; SUMO-3; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B
Gene Symbols SUMO3
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the human SUMO-3 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6612
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.