missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.
- Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
- Space saving: Products require only one-tenth of the space required for ready-made solutions
- Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate
Specifications
Specifications
| Color | Colorless |
| CAS | 7664-93-9 |
| Boiling Point | 100°C |
| Concentration | 0.05mol/L |
| Density | 1.1g/cm3 at 20°C |
| Packaging | LDPE ampule |
| Vapor Pressure | 1.100 hPa (50°C) |
| Normality | 0.1N |
| Quantity | For dilution to 1 L |
| Molecular Formula | H2O4S |
| Show More |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?