missing translation for 'onlineSavingsMsg'
Learn More

Styrene-d8, for NMR, 98+ atom% D, stabilized

Product Code. 10125113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Ampoule
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10125113 1 g Ampoule 1g
10072360 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10125113 Supplier Thermo Scientific Acros Supplier No. 175390010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 19361-62-7 | C8H8 | 112.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19361-62-7
Molecular Formula C8H8
Molecular Weight (g/mol) 112.201
MDL Number MFCD00044231
InChI Key PPBRXRYQALVLMV-GDALLCCDSA-N
Synonym styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
PubChem CID 88025
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
SMILES C=CC1=CC=CC=C1

Specifications

Physical Form Liquid
Color Colorless
Density 0.9700g/mL
Deuterium (D) 98.0% min.
Flash Point 32°C
Packaging Ampoule
Refractive Index 1.5443
Water 0.05% max. (Coulometric)
Quantity 1 g
Boiling Point 50.0°C to 51.0°C (25.0mmHg)
Linear Formula C6D5CD=CD2
Formula Weight 112.22
Grade NMR
Chemical Name or Material Styrene-d8, 98+ atom% D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Styrene-d8, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains 4-tert-Butyl catechol. May produce an allergic reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.