missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Styragel GPC Column
Description
- Styragel HMW columns are packed with a 20 μm particle and utilize a special 10 μm frit to minimize polymer shearing, making them ideal for use in high-molecular weight applications.
Specifications
Specifications
| Diameter (Metric) | 7.8 mm |
| Length (Metric) | 300 mm |
| Particle Shape | Spherical |
| Particle Size | 15 to 20 μm |
| Pore Size | 7 to 10 Å |
| Quantity | 1/Ea. |
| Type | Column |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?