missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STRN4 Recombinant Rabbit Monoclonal Antibody (K01_1Y83)
GREENER_CHOICE

Product Code. 30829371
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829371 100 μL
30828078 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829371 Supplier Invitrogen™ Supplier No. MA563781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

STRN4 Recombinant Monoclonal Antibody for Western Blot

Binds calmodulin in a calcium dependent manner. May function as scaffolding or signaling protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRN4
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Y83
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Strn4
Gene Accession No. Q9NRL3
Gene Alias C80611; PPP2R6C; protein phosphatase 2 regulatory subunit B'''gamma; striatin 4; striatin, calmodulin binding protein 4; striatin-4; Strn4; Zin; zinedin
Gene Symbols Strn4
Host Species Rabbit
Immunogen Recombinant protein of human Striatin 4.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29888
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.