missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STRN3 Monoclonal Antibody (S68)
GREENER_CHOICE

Product Code. 10100924
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10100924 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10100924 Supplier Invitrogen™ Supplier No. MA146461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody is specific for SG2NA. It does not cross-react with Striatin.

SG2NA was identified as a human cancer autoantigen and has now been implicated in vesicular trafficking and signal transduction. It is highly related to striatin and zinedin. It is a scaffolding protein that binds PP2A A and C subunits as well as the human homolog of the yeast protein, Mob1, which has been implicated in control of mitotic progression in yeast. It also binds calmodulin and appears to modulate PP2A activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRN3
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone S68
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.02% sodium azide
Gene STRN3
Gene Accession No. P58405, Q13033, Q9ERG2
Gene Alias Cell cycle autoantigen SG2NA; cell cycle S/G2 nuclear autoantigen; cell-cycle autoantigen SG2NA; Gs2na; nuclear autoantigen; PPP2R6B; protein phosphatase 2 regulatory subunit B'''beta; s/G2 antigen; Sg2na; SG2NA beta isoform; striatin 3; striatin, calmodulin binding protein 3; striatin-3; striatin-3 35 kDa; striatin-3-gamma; Strn3
Gene Symbols STRN3
Host Species Mouse
Immunogen Synthetic peptide made to an internal portion of human SG2NA (within residues 200-300).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114520, 29966, 94186
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.