missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Streptavidin, Pacific Orange™ conjugate
Description
Pacific Orange™ streptavidin has absorption/emission maxima of ∼400/551 nm. Streptavidin is a nonglycosylated biotin-binding protein that is isolated from bacteria and has a near-neutral isoelectric point. It is widely used to detect biotinylated probes, because it reportedly exhibits less nonspecific binding than does the glycosylated, biotin-binding protein, avidin.
View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
Specifications
Specifications
| Conjugate | Pacific Orange |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis |
| Quantity | 1 mg |
| Content And Storage | Contains 1 vial of streptavidin Pacific Orange™ conjugate. Store in freezer (-5 to -30°C) and protect from light. |
| Excitation/Emission | 400/551 |
| Flow Cytometer Laser Lines | 405 |
| Shipping Condition | Room Temperature |
| Form | Solid |
| Product Line | Pacific Orange |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?