missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Streptavidin HRP
Description
Purified from Streptomyces avidinii and provided in a phosphate buffered saline solution. The streptavidin/avidin-biotin staining and detection strategy is used when increased assay sensitivity is desired for a wide variety of systems.
Format/Formulation: HRP Conjugate
Format/Formulation: HRP Conjugate
Specifications
Specifications
| Concentration | 2 mg/mL |
| Conjugate | HRP |
| Quantity | 1 mg |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Form | Liquid |
| Product Type | Streptavidin Conjugate (fluorescent) |
| Label or Dye | HRP (Horseradish Peroxidase) |
For Laboratory Use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?