missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin, Allophycocyanin, crosslinked, conjugate
Description
Streptavidin is a bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin. Allophycocyanin is an intensely bright phycobiliprotein with a broad absorption profile that peaks at ∼650 nm, and a fluorescence emission maximum of ∼660 nm.
View a selection guide for all streptavidin RPE, APC, and tandem conjugates.
View a selection guide for all streptavidin RPE, APC, and tandem conjugates.
Specifications
Specifications
| Concentration | 1 mg/mL |
| Conjugate | APC |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis |
| Quantity | 0.5 mL |
| Content And Storage | Contains 1 vial of streptavidin APC , crosslinked, conjugate (1 mg/mL). Store in refrigerator (2–8°C) and protect from light. |
| Excitation/Emission | 650/660 |
| Flow Cytometer Laser Lines | 633 or 635 |
| Shipping Condition | Wet Ice |
| Form | Solution |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?