Learn More
Lenz Laborglasintrumente Stopcocks, Straight Bore, with Solid Glass Plug
Description
- Acc. to DIN 12541-1
- With solid glass plug and screwthread retaining device
- One straight bore
- Material: borosilicate glass 3.3
- Leakage rate acc. to DIN 12540-1
Specifications
Specifications
| Plug Bore | 2.5 mm |
| Material | Borosilicate Glass (3.3) |