Learn More
Lenz Laborglasintrumente Stirrer with Stirrer Bearing
Description
- Shaft with blade and PTFE security screw
- Gastight closure of vacuum stirring apparatuses up to 10-3 mbar at a continuous operation speed of max.
- 800 rpm, for round bottom flasks 250-2000 mL
- Shaft made of DURAN™ tubing
Specifications
Specifications
| Description | Screw cap, GL 25 |
| For Use With (Equipment) | For round bottom flasks 250 to 2000 mL |
| Material | Polypropylene |