Learn More
KARTELL Sterile Petri Dish
Product Code. 10051001
Change view
Click to view available options
Diameter (Metric):
60 mm
120 mm
Unit Size:
Pack of 1000
Pack of 320
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10051001 120 mm Pack of 320
16697872 60 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10051001 Supplier KARTELL Supplier No. 0036100
Only null left
Add to basket
Add to basket
The Sterile Petri Dish is a disposable planar, clear Petri dish designed for routine laboratory work.
  • Disposable planar, clear Petri dishes in PS.
  • Ventilated and radiation- sterilized.
  • Designed for routine lab work, bacteriological research.
  • CE Marked.
  • With aeration notches on the lid to prevent condensation formation.
  • Beta-ray sterilization
  • Designed for routine lab work, bacteriological research.
  • Gas exchange outside Petri dish is reduced thanks to the absence of notches.
Specifications
Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Disposable Single-use
Gridded Not Gridded
Material Polystyrene
Shape Circular
Sterility Sterile
Surface Treatment Non-treated
Type Petri Dish
Well Count 1
Height (Metric) 14 mm
Diameter (Metric) 120 mm