missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAT2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10144503
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10144503 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10144503 Supplier Invitrogen™ Supplier No. 44362G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

STAT2 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, IP, ChIP

STAT2 is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. In response to interferon (IFN), this protein forms a complex with STAT1 and IFN regulatory factor family protein p48 (ISGF3G), in which this protein acts as a transactivator, but lacks the ability to bind DNA directly. Transcription adaptor P300/CBP (EP300/CREBBP) has been shown to interact specifically with this protein, which is thought to be involved in the process of blocking IFN-alpha response by adenovirus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene Stat2
Gene Accession No. P52630
Gene Alias 1110034C02Rik; 1600010G07Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; Aprf; AW109958; AW496480; DNA-binding protein APRF; EGK_03803; HIES; IMD44; interferon alpha induced transcriptional activator; ISGF-3; MGC59816; p113; signal transducer and activator of transcription 2; signal transducer and activator of transcription 2, 113kDa; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); STAT; STAT 2; STAT113; STAT2; STAT-2; Stat3
Gene Symbols Stat2
Host Species Rabbit
Immunogen The antiserum was produced against a chemically synthesized peptide derived from the C-terminal region of human STAT2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6773
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.