missing translation for 'onlineSavingsMsg'
Learn More

BOLA Star Head Magnetic Stirring Bars

Product Code. 10174531
Change view
Click to view available options
Diameter (Metric):
35 mm
40 mm
58 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10174531 35 mm Each
10594012 40 mm Each
10629021 58 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10174531 Supplier BOLA Supplier No. C36019
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Star Head Magnetic Stirring Bars PTFE-coated

PTFE-encapsulated magnetic core (Alnico 5), universal chemical resistance.

Specifications

Autoclavable Autoclavable
Color White
Height (Metric) 12 mm
Coating PTFE
Diameter (Metric) 35 mm
Shape Star
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.