Learn More
RETSCH Stainless Steel Intermediate Pan for Test Sieves
Product Code. 10157221 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric) Outer:
200 mm
203 mm
305 mm
400 mm
450 mm
Height (Metric):
25 mm
25.4 mm
40 mm
50 mm
50.8 mm
65 mm
100 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Height (Metric) unitSize
10157221 203 mm 50.8 mm Each
11954432 450 mm 100 mm Each
10147221 200 mm 25 mm Each
11974422 203 mm 25.4 mm Each
11904432 305 mm 40 mm Each
10544692 200 mm 50 mm Each
11914432 400 mm 65 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10157221 Supplier RETSCH Supplier No. 69.220.3050
Only null left
Add to basket
Add to basket
Collects resulting samples from test sieves. These Stainless Steel intermediate pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter.
  • Stainless Steel
  • Compatible with multiple test sieves, based upon diameter
  • Excellent product quality due to extensive optical inspection
  • Sieve frame with a solid base
Specifications
Type Intermediate Pan
Material Stainless Steel
For Use With (Equipment) Test sieves; Vibratory Sieve Shaker AS 200 basic
Height (Metric) 50.8 mm
Diameter (Metric) Outer 203 mm