missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Stainless Steel Forceps Smooth Jaws

Product Code. 17682975 Shop All Nickel-Electro Products
Change view
Click to view available options
Length (Metric):
115 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
17682975 115 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17682975 Supplier Nickel Electro™ Supplier No. 7101SJ
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory stainless steel forceps with smooth jaws.

  • Laboratory forceps
  • Stainless steel
  • Smooth Jaws
  • 115 mm
  • Sharp ends

Specifications

Tip Style Sharp, Straight
Length (English) 6 in.
Material Stainless Steel
Length (Metric) 115 mm
Product Type Forceps
Sterility Non-sterile
Style Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.