Learn More
Gilson™ Stainless Steel Connectors
Product Code. 10444522 Shop All Gilson Products
Change view
Click to view available options
Diameter (Metric):
0.6 mm
0.8 mm
1.1 mm
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10444522 0.8 mm Pack of 5
10434522 0.6 mm Pack of 5
10183092 1.1 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10444522 Supplier Gilson™ Supplier No. F117986
Only null left
Add to basket
Add to basket
Metal sleeve connectors for small bore tubing
  • Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
  • ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
  • PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
  • PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 0.8 mm
Material Stainless Steel
Packaging Quantity 5 Set
Includes Metal sleeve
Product Type Connector