Learn More
Bochem™ Stainless Steel Clamp
Description
- 18/10 stainless steel
- Round jaws
- DIN 12894
Specifications
Specifications
| Quantity | 1 Ea. |
| Diameter (Metric) | 60 mm |
| Length (Metric) Arm | 150 mm |
| Jaw Type | Round |