Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F1, NVLAP
Product Code. 16672442 Shop All Ohaus Products
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
Unit Size:
Each
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass unitSize
16672442 1 kg Each
16692422 1 mg Each
16602432 2 mg Each
16612432 5 mg Each
16622432 10 mg Each
16632432 20 mg Each
16642432 50 mg Each
16652432 100 mg Each
16662432 200 mg Each
16672432 500 mg Each
16682432 1 g Each
16692432 2 g Each
16602442 5 g Each
16612442 10 g Each
16622442 20 g Each
16632442 50 g Each
16642442 100 g Each
16652442 200 g Each
16662442 500 g Each
16682442 2 kg Each
16692442 5 kg Each
16602452 10 kg Each
16612452 20 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Product Code. 16672442 Supplier Ohaus™ Supplier No. 80780349
Only null left
Add to basket
Add to basket
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F1 Tolerances
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NVLAP Accredited Certificate
Material Troemner Alloy 8
Weight (Metric) 1.500 kg
Mass 1 kg
Tolerance 5 mg
Includes Protective Case