missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SR Monoclonal Antibody (16H3 (16H3E8))
GREENER_CHOICE

Product Code. 10057612
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10057612 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10057612 Supplier Invitrogen™ Supplier No. 339300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

This monoclonal antibody can be used to detect a sub-set of the non-snRNP splicing factors termed SR proteins. The antibody detects SRp75, SRp55, SRp40, and SRp20 but not SRp30a or b (ASF/SF2, SC-35) proteins. 16H3 also recognizes approximately 20 distinct nuclear proteins including the U1 70K component of the U1 snRNP and both subunits of U2AF. The epitope detected by the 16H3 monoclonal antibody has been termed the alternating arginine domain and is composed almost exclusively of argnine alternating with glutamate and aspartate. 16H3 targets are predominantly nuclear, nonnucleolar, and are localized to active sites of polymerase II transcription.

SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear speckles, in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 16H3 (16H3E8)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene SRSF6
Gene Accession No. G3V6S8, O18776, O35326, P84103, P84104, Q08170, Q09167, Q13243, Q13247, Q3B7L6, Q3SZR8, Q3TWW8, Q8VE97
Gene Alias 1210001E11Rik; 5730499P16Rik; AI314910; AL024116; AW146126; AW550192; B52; Cl-4; Delayed-early protein HRS; epididymis secretory protein Li 91; HEL-S-91; HRS; hypothetical protein; insulin-induced growth response protein CL-4; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 4; MNCb-2616; pre-mRNA splicing factor SRp20; pre-mRNA-splicing factor SRP20; Pre-mRNA-splicing factor SRP40; pre-mRNA-splicing factor SRP55; Pre-mRNA-splicing factor SRP75; Protein X16; RGD1561347; serine and arginine rich splicing factor 3; serine and arginine rich splicing factor 4; serine and arginine rich splicing factor 5; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 3; Serine/arginine-rich splicing factor 4; serine/arginine-rich splicing factor 5; Serine/arginine-rich splicing factor 6; SFRS3; Sfrs4; Sfrs5; Sfrs6; splicing factor; splicing factor arginine/serine-rich 5 (SRp40 HRS); splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 20-kD; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 4; SR splicing factor 5; SR splicing factor 6; SRP001LB; Srp20; SRP40; SRP55; SRP55-1; SRP75; SRSF3; SRSF4; SRSF5; Srsf6; X16
Gene Symbols SRSF3, SRSF4, SRSF5, SRSF6
Host Species Mouse
Immunogen Purified, de-phosphorylated bovine Srp55 and Drosophila p55-GST. The 16H3 epitope has been mapped to a 40-amino acid polypeptide composed almost exclusively of arginine alternating with glutamate and aspartate.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008654, 100340192, 100347748, 100350179, 20383, 20384, 29667, 361814, 362264, 362612, 403667, 403687, 419116, 419600, 419815, 423258, 478160, 490754, 507828, 510474, 540276, 57317, 614310, 6428, 6429, 6430, 6431, 67996
Target Species Human, Mouse, Rat, Canine, Rabbit, Bovine, Chicken, Drosophila, Frog
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.