missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 12813

Description
SR 12813 is a pregnane X receptor (PXR) agonist (EC50 values are 200 and 700 nM for human and rabbit PXR respectively). Activates the farnesoid X receptor (FXR) at μM concentrations. Displays hypocholesterolemic activity via enhanced degradation of HMG-CoA reductase.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 504.53 |
| Purity | >99% |
| Molecular Weight (g/mol) | 504.541 |
| Chemical Name or Material | SR 12813 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?