missing translation for 'onlineSavingsMsg'
Learn More

Squalamine lactate, MedChemExpress

Product Code. 16493097
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493097 50 mg 50mg
16447058 10 mg 10mg
16421619 1 mg 1mg
16409607 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493097 Supplier Medchem Express Supplier No. HY16467

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Squalamine lactate is an aminosterol compound discovered in the tissues of the dogfish shark, with antimicrobial activity, and used for the treatment of neovascular age-related macular degeneration.

Squalamine lactate is an aminosterol compound discovered in the tissues of the dogfish shark, with antimicrobial activity, and used for the treatment of neovascular age-related macular degeneration.

Specifications

Chemical Name or Material Squalamine lactate
Physical Form Solid
CAS 320725-47-1
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C37H71N3O8S
For Use With (Application) COVID-19-immunoregulation
Synonym MSI-1256F
Solubility Information DMSO : ≥ 36.66 mg/mL (51.06 mM)
SMILES NCCCCNCCCN[C@H]1CC[C@@]2(C)[C@](C[C@@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CC[C@@H](OS(=O)(O)=O)C(C)C)([H])C1.C[C@@H](C(O)=O)O
Molecular Weight (g/mol) 718.04
Formula Weight 718.04
Percent Purity 98.37%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.