missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Solution dNTPs

Product Code. 10024684 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
dATP
dCTP
dGTP
dTTP
dUTP
Quantity:
25 μm
100 μm
500 μm
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
10024684 dUTP 25 μm Each
10238084 dATP 25 μm Each
10545335 dGTP 25 μm Each
10389764 dGTP 100 μm Each
10648035 dCTP 25 μm Each
10430125 dCTP 100 μm Each
10527065 dTTP 100 μm Each
10074294 dTTP 25 μm Each
10526875 dGTP 500 μm Each
10004824 dATP 100 μm Each
10665355 dUTP 100 μm Each
10537065 dTTP 500 μm Each
10350055 dATP 500 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 10024684 Supplier Cytiva Supplier No. 28406541

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use dNTPs incorporating the most recent advances in manufacturing technology. Cytiva illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

 

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

 

Order Info

Custom formulation, testing, packaging, and labeling are available

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7
Product Type dUTP
Concentration 100 mM
Purity >99%
Quantity 25 μm
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.