missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Solution dNTP Sets

Product Code. 10004304 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4 x 25 μm
4 x 100 μm
4 x 500 μm
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10004304 4 x 500 μm Pack of 4
10084724 4 x 25 μm Pack of 4
10267704 4 x 100 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10004304 Supplier Cytiva Supplier No. 28406553

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use dNTP sets for optimum performance and consistency. Cytiva Lifescience™ illustra™ Solution dNTP Sets are free from DNase, RNase, and nicking enzyme activity.

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7
Product Type dNTP set
Concentration 100 mM
Purity >99%
Quantity 4 x 500 μm
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.