missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Cream or Tan |
| Assay Percent Range | 95% |
| Packaging | Glass bottle |
| Linear Formula | CH3CH2C(CH3)2ONa |
| Quantity | 500 g |
| Formula Weight | 110.13 |
| Percent Purity | 95% |
| Grade | Pure |
| Physical Form | Powder |
| Chemical Name or Material | Sodium tert-pentoxide, 95% |
Safety and Handling
Product Identifier
- Sodium tert-pentoxide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Self-heating substance Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H251-Self-heating: may catch fire.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?