missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide Hydrate, Honeywell™

Product Code. p-7120830 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15691440 250 g Glass Bottle 250g
15681440 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15691440 Supplier Honeywell Research Chemicals Supplier No. 14738250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

for gravimetric analysis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
PubChem CID 45051681
SMILES [Na+].[Na+].[S--]

Specifications

Color Colorless
Quantity 250 g
UN Number UN1849
IUPAC Name disodium;sulfanide;hydrate
Formula Weight 78.04g/mol (anhydrous basis)
Percent Purity 59 to 65%
Packaging Glass Bottle
Appearance Flakes
Chemical Name or Material Sodium Sulfide Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.