missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions

Product Code. 10185753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10185753 2.5 kg Plastic Bottle 2.5kg
10782602 500 g Plastic Bottle 500g
15139153 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10185753 Supplier Thermo Scientific Acros Supplier No. 424370025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3 (5% soln. at 25°C)
Physical Form Granular Powder
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula Na2HPO4
Merck Index 15, 8789
Solubility Information Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 0.2% max. (105°C)
Packaging Plastic Bottle
Decomposition Information Decomposes information: > 240°C
Chemical Name or Material Sodium phosphate, dibasic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.