missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, extra pure, 50 wt% solution in water

Product Code. 10175150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
5 L
25 L
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
1 L
2.5 L
25 L
25mL
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175150 25 mL Plastic Bottle 25mL
10771974 1 L Plastic Bottle 1 L
10781321 2.5 L Plastic Bottle 2.5 L
10544901 5 L Plastic drum 5 L
10043313 25 L Plastic drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10175150 Supplier Thermo Scientific Acros Supplier No. 259860250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-73-2 | HNaO | 39.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color White to Gray
Boiling Point 145.0°C
Melting Point 12.0°C
Linear Formula NaOH
UN Number 1824
Fieser 01,1083; 05,616; 07,336; 08,460
Density 1.5000g/mL
Merck Index 15, 8761
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone and ether
Quantity 25 mL
Formula Weight 40
Percent Purity 49.0 to 51.0% (Acidimetry)
Grade Extra Pure
Packaging Plastic Bottle
Specific Gravity 1.5
Physical Form Powder
Chemical Name or Material Sodium hydroxide, 50 wt% solution in water
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide, 50 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.