missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate, Honeywell Fluka™

Product Code. p-7071836 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15651580 50 g 50g
15641580 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15651580 Supplier Honeywell-Fluka Supplier No. 1784150G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for HPLC,≥99.0% (NT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.007
MDL Number MFCD00013101
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Quantity 50 g
Linear Formula HCOONa
UN Number NONH for all modes of transport
Beilstein 3595134
Solubility Information Soluble in water 0.1g/mL at 25°C
Formula Weight 68.01g/mol
Percent Purity ≥99% (NT)
Chemical Name or Material Sodium Formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.