missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, 99.5%, for analysis

Product Code. 10115793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10115793 2.5 kg Plastic bottle 2.5kg
10107903 100 g Plastic bottle 100g
10174620 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10115793 Supplier Thermo Scientific Acros Supplier No. 246900025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-15-6 | BrNa | 102.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390.0°C
Melting Point 755.0°C
Color White
pH 5 to 8.8 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99.5% min. (Argentometry)
Linear Formula NaBr
Fieser 12,445; 13,46
Merck Index 15, 8729
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C),1210g/L water (100°C),1g/6 mL methanol,7g/100 mL 95% ethanol
IUPAC Name sodium bromide
Formula Weight 102.89
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium bromide, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.