missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfite, ACS reagent, powder

Product Code. 10140862 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
2.5kg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10140862 2.5 kg Plastic bottle 2.5kg
10697581 5 g Glass bottle 5g
10552072 100 g Plastic bottle 100g
10474511 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10140862 Supplier Thermo Scientific Acros Supplier No. 419440025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7631-90-5 | NaO3S | 103.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
PubChem CID 23665763
ChEBI CHEBI:26709
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Melting Point 150.0°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Linear Formula NaHSO3
Fieser 01,1047; 11,487; 16,272
Merck Index 15, 8724
Solubility Information Solubility in water: 300g/L. Other solubilities: soluble in 2 parts boiling water, soluble in about 70 parts alcohol
Formula Weight 104.06
Percent Purity ≥58.5% (SO2)
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 1.48
Density 1.4800g/mL
Chemical Name or Material Sodium bisulfite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.