missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals

Product Code. 10147850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147850 500 g Glass bottle 500g
10288513 2.5 kg Glass bottle 2.5kg
15149153 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147850 Supplier Thermo Scientific Acros Supplier No. 419435000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
IUPAC Name sodium;hydrogen sulfate;hydrate
SMILES O.[Na+].OS([O-])(=O)=O

Specifikationer

Melting Point 58.0°C
Color White
Physical Form Adhering Crystals
Quantity 500 g
Assay Percent Range 97%
Linear Formula NaHSO4·H2O
Merck Index 15,8722
Solubility Information Solubility in water: 670g/L. Other solubilities: decomposes by alcohol into sodium sulfate and, h2so4
Formula Weight 138.08
Percent Purity 97%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Sodium bisulfate monohydrate, 97%
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Sodium bisulfate monohydrate
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
Faroangivelser
  • H318-Causes serious eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.