missing translation for 'onlineSavingsMsg'
Learn More

Sodium benzoate, 99+%, extra pure

Product Code. 10063903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10063903 5 kg Plastic Drum 5kg
10467540 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10063903 Supplier Thermo Scientific Acros Supplier No. 148980050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
PubChem CID 517055
IUPAC Name sodium;benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Assay Percent Range 99+%
Linear Formula C6H5CO2Na
Beilstein 09, 92
Fieser 01,1044; 02,377
Merck Index 15, 8718
Formula Weight 144.11
Percent Purity ≥99.0% (on dried substance)
Grade Extra Pure
Quantity 5 kg
Infrared Spectrum Authentic
Packaging Plastic Drum
Physical Form Crystals, Granules, Flakes or Crystalline Powder
Chemical Name or Material Sodium benzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.