missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep™
Shop All SPEX Certiprep ProductsDescription
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ag | 1000μg/mL |
Specifications
Specifications
| Packaging | Bottle |
| Certificate of Analysis | Assurance Certificate of Analysis reports up to 30 elements at ppb levels. |
| Technique | AA, ICP |
| Type | Certified Reference Material |
| IUPAC Name | silver(1+) nitrate |
| Quantity | 250 mL |
| Chemical Name or Material | Silver Standard |
Safety and Handling
Product Identifier
- 1000 ug/mL Silver
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: nitric acid, silvernitrate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?