missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrite, 99%

Product Code. p-3756701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10790711 10 g Glass bottle 10g
10772305 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10790711 Supplier Thermo Scientific Acros Supplier No. 210690100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7783-99-5 | AgNO2 | 153.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-99-5
Molecular Formula AgNO2
Molecular Weight (g/mol) 153.88
MDL Number MFCD00003413
InChI Key KKKDGYXNGYJJRX-UHFFFAOYSA-M
Synonym silver nitrite, unii-t3mz57ogif, t3mz57ogif, silbernitrit, nitrous acid, silver 1+ salt, silver i nitrite, silvernitrite, acmc-20ajq7, ag.no2, silver nitrite 10g
PubChem CID 160904
IUPAC Name silver;nitrite
SMILES N(=O)[O-].[Ag+]

Specifications

Melting Point 140.0°C
Color Yellow to Green
Physical Form Powder and/or Agglomerates
Quantity 10 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula AgNO2
Fieser 01,1011; 06,515; 11,467
Merck Index 15, 8658
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: decomposes in diluted acids, insoluble in alcohol
Formula Weight 153.88
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 4.45
Density 4.4500g/mL
Chemical Name or Material Silver nitrite
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Silver nitrite
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.