missing translation for 'onlineSavingsMsg'
Learn More

Draeger Safety Silica Gel Tube

Product Code. 10180821
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10180821 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10180821 Supplier Draeger Safety Supplier No. 6728851

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Draeger Safety Silica Gel Tube is specially suitable for the sampling of organic compounds which are present in high concentrations in the air sample.

  • For example the analysis of exhaust air for the determination of the emission of a harmful substance
  • After sampling the tube must be sealed with the polyethylene supplied caps
  • Adsorbate: organic compounds which adsorb on silicagel
  • Adsorption layer: 1100 mg

Specifications

Description 125 mm tube length, 7 mm O.D., 5 mm I.D.
Type Type G
Quantity 10/Pk.
Tube Type Silica Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.