missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Validated siRNA

Product Code. 10085064
Change view
Click to view available options
Purity:
CY3 Labeled
FAM Labeled
HPL-OLIGOC
HPLC
In Vivo
Standard
Quantity:
1 μmol
10 μmol
20 nmol
250 nmol
40 nmol
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Purity Quantity unitSize
10085064 In Vivo 10 μmol Each
10534075 Standard 40 nmol Each
10724047 HPLC 40 nmol Each
10177964 HPL-OLIGOC 250 nmol Each
10515005 In Vivo 1 μmol Each
10289494 In Vivo 250 nmol Each
10724237 CY3 Labeled 20 nmol Each
10734237 FAM Labeled 20 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10085064 Supplier Ambion™ Supplier No. AM51343

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.

  • The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry
  • Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression
  • Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening
  • The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment

The benefits of using Silencer™ Select siRNAs include:

  • Potent – up to 100-fold more potent than currently available siRNAs and fewer off-target effects
  • Most Specific – LNA™ chemical modifications reduce off-target effects by up to 90%
  • Reliable – demonstrated improvements in consistency and reliability of phenotypic results
  • Guaranteed – 100% guaranteed to silence, the best guarantee in the industry

Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.

Approximate Turnaround Times*

  • Standard purification: 4 business days
  • HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
  • In Vivo Ready: 15 business days (250 nmol)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Silencer™ Validated siRNA
RNAi Type siRNA
Product Line Silencer
Purity In Vivo
Quantity 10 μmol
Temperature (Metric) Store at room temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.