Learn More
Epredia™ Signature Series™ Pen-Fix™
Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.
Brand:  Epredia™ 6101
Code : A0
Additional Details : CAS Number : 64-17-5 Weight : 12.62000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
 View more versions of this product
Product Code. 10148438
  • £78.39 / Pack of 4
Estimated Shipment: 26-04-2022
Add to basket
Description
Description
  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative
Specifications
Specifications
4 x 1 gal.
Videos
SDS
Documents
Certificates
View accessory products