Learn More
Bochem™ Sharp Forceps with Guide-pin
Product Code. 10468242
Change view
Click to view available options
Length (Metric):
115 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10468242 115 mm Each
1 options
Product Code. 10468242 Supplier Bochem™ Supplier No. 1131
Only null left
Add to basket
Add to basket
Sharply pointed, straight forceps with a guide-pin. Bochem™ Sharp Forceps with Guide-pin are constructed of stainless steel.
  • Stainless steel construction.
  • Straight, sharp point design.
  • Guide-pin.
Specifications
Material Stainless Steel
Length (Metric) 115 mm
Product Type Forceps
Style Sharp, straight