missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Bluing Reagent

Product Code. 10127998 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1L
4L
Unit Size:
4 L
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10127998 4L 4 L
10381775 1L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10127998 Supplier Epredia™ Supplier No. 6769002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Employ a gentle, pH-controlled solution for bluing hematoxylin with the Epredia™ Shandon™ Bluing Reagent.

  • Ready-to-use
  • Thoroughly pH-controlled for completely effective bluing
TRUSTED_SUSTAINABILITY

Specifications

Type Bluing Reagent
Quantity 4L
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 1
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H370-Causes damage to organs.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P271-Use only outdoors or in a well-ventilated area.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P235-Keep cool.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: sodium bicarbonate,lithium carbonate,methyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.