missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SH2D1A Polyclonal Antibody

Product Code. 10142084
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10142084 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10142084 Supplier Invitrogen™ Supplier No. PA112645

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control for this product is NK-92 cell lysate.

This gene encodes a protein that plays a major role in the bidirectional stimulation of T and B cells. This protein contains an SH2 domain and a short tail. It associates with the signaling lymphocyte-activation molecule, thereby acting as an inhibitor of this transmembrane protein by blocking the recruitment of the SH2-domain-containing signal-transduction molecule SHP-2 to its docking site. This protein can also bind to other related surface molecules that are expressed on activated T, B and NK cells, thereby modifying signal transduction pathways in these cells. Mutations in this gene cause lymphoproliferative syndrome X-linked type 1 or Duncan disease, a rare immunodeficiency characterized by extreme susceptibility to infection with Epstein-Barr virus, with symptoms including severe mononucleosis and malignant lymphoma. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D1A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene Sh2d1a
Gene Accession No. O60880
Gene Alias DSHP; Duncan disease homolog; Duncan disease SH2-protein; Duncan's disease; EBVS; FLJ18687; FLJ92177; Gm686; HGNC:10820; IMD5; LYP; MTCP1; RGD1562408; RP5-1052M9.3; SAP; SAP/SH2D1A; SH2 domain containing 1A; SH2 domain protein 1A; SH2 domain-containing protein 1A; SH2D1A; signaling lymphocyte activation molecule-associated protein; signaling lymphocytic activation molecule-associated protein; SLAM associated protein/SH2 domain protein 1A; SLAM-associated protein; SLAM-associated protein isoform SAP-2; T cell signal transduction molecule SAP; T-cell signal transduction molecule SAP; Xlp; XLPD; XLPD1
Gene Symbols Sh2d1a
Host Species Rabbit
Immunogen Full-length recombinant SAP protein.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4068
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.