Learn More
Trajan™ VSV-6 Vespel Valve Seat
Product Code. 10114123
Change view
Click to view available options
For Use With (Equipment):
Replacement for VSV-6 Micro Valves
Unit Size:
Pack of 5
Product Code. For Use With (Equipment) unitSize
10114123 Replacement for VSV-6 Micro Valves Pack of 5
1 options
Product Code. 10114123 Supplier Trajan™ Supplier No. 123681
Precision metering and on-off control of gas flows. Designed specifically for chromatography applications and may be used at elevated temperatures from high vacuum to medium pressure.
  • Ideal for chromatography applications
  • May be used at elevated temperatures from high vacuum to medium pressure
  • Vacuum rated to a leak rate of 1E-05 cc/sec of helium
Specifications
Type Valve Seat, Vespel
Style VSV-6 SGE
For Use With (Equipment) Replacement for VSV-6 Micro Valves