missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SETD8 Monoclonal Antibody (43AT890)
GREENER_CHOICE

Product Code. 10111080
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10111080 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10111080 Supplier Invitrogen™ Supplier No. MA511141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SETD8 Monoclonal Antibody for Western Blot

SETD8 is a histone methyltransferase that specifically monomethylates 'Lys-20' of histone H4. H4 'Lys-20' monomethylation is enriched during mitosis and represents a specific tag for epigenetic transcriptional repression. The protein mainly functions in euchromatin regions, thereby playing a central role in the silencing of euchromatic genes. It is required for cell proliferation, probably by contributing to the maintenance of proper higher order structure of DNA during mitosis. It is also involved in chromosome condensation and proper cytokinesis. Nucleosomes are preferred as substrate compared to free histones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD8
Applications Western Blot
Classification Monoclonal
Clone 43AT890
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene KMT5A
Gene Accession No. Q2YDW7, Q9NQR1
Gene Alias 2410195B05Rik; AA617402; AW536475; H4-K20-HMTase; H4-K20-HMTase KMT5A; H4-K20-HMTase SETD8; H4-K20-specific histone methyltransferase; H4K20-specific histone methyltransferase splice variant Set8b; histone methyltransferase Pr-set7/Set8; histone-lysine N-methyltransferase KMT5A; histone-lysine N-methyltransferase SETD8; KMT5A; lysine (K)-specific methyltransferase 5A; lysine methyltransferase 5A; lysine N-methyltransferase 5A; Lysine-specific methylase 5A; N-lysine methyltransferase KMT5A; N-lysine methyltransferase SETD8; PR/SET domain containing protein 8; PR/SET domain-containing protein 07; PR/SET07; PRSET7; PR-Set7; RGD1305893; SET domain containing (lysine methyltransferase) 8; SET domain containing protein 8 variant 1; SET domain containing protein 8 variant 2; SET domain-containing 8; SET domain-containing protein 8; SET07; SET8; SETD8; Unknown (protein for MGC:134426)
Gene Symbols KMT5A
Host Species Mouse
Immunogen Purified recombinant GST fusion protein encoding N-terminal of human SET07.
Purification Method Size-exclusion - Dialysis, Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387893, 67956
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.