missing translation for 'onlineSavingsMsg'
Learn More

Kartell SpA Spare Parts for Pluripet

Product Code. 10162362
Change view
Click to view available options
Volume (Metric):
10 μL
10,000 μL
100 μL
1000 μL
2 μL
20 μL
200 μL
5000 μL
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10162362 5000 μL Each
10663103 2 μL Each
10425602 10 μL Each
10376192 20 μL Each
10435602 100 μL Each
10152362 200 μL Each
10560623 1000 μL Each
10466362 10,000 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10162362 Supplier Kartell SpA Supplier No. 0134600
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Spare part set for Pluripet including: 1 cone, 2 PTFE seal, 2 O-ring, 1 cone nut

Spare part set for Pluripet

Includes:

  • 1 cone
  • 2 PTFE seal
  • 2 O-ring
  • 1 cone nut

Specifications

Autoclavable Autoclavable
Color Assorted
Disposable Reusable
For Use With Manual Air Displacement Pipette
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Sterility Non-sterile
Type Pipette Kit
Volume (Metric) 5000 μL
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.