missing translation for 'onlineSavingsMsg'
Learn More

Kartell SpA Spare Parts for Fixopet

Product Code. 10192552
Change view
Click to view available options
Volume (Metric):
10 μL
50 μL
100 μL
200 μL
500 μL
1000 μL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10192552 200 μL Each
10192502 10 μL Each
10487332 50 μL Each
10476362 100 μL Each
10500823 500 μL Each
10501013 1000 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10192552 Supplier Kartell SpA Supplier No. 0135400
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Spare part set for Fixopet

Spare part set for Fixopet

Includes:

  • 1 cone
  • 2 PTFE seal
  • 2 O-ring
  • 1 cone nut

Specifications

Autoclavable Autoclavable
Color Assorted
Disposable Reusable
For Use With Manual Air Displacement Pipette
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Sterility Non-sterile
Type Pipette Kit
Volume (Metric) 200 μL
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.