missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequenase Version 2.0 DNA Polymerase

Product Code. 15819896
Change view
Click to view available options
Quantity:
1000 U
200 U
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15819896 1000 U Each
15809896 200 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 15819896 Supplier Applied Biosystems™ Supplier No. 70775Z1000UN

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Sequenase™ Version 2.0 DNA Polymerase is a genetically engineered form of T7 DNA polymerase. Unlike the wild-type enzyme it has virtually no 3'→5' exonuclease activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 13 units/μL
Description Sequenase Version 2.0 DNA Polymerase, 1000 Units, 13 units/μL Concentration, Genetically-Engineered form of T7 DNA Polymerase, 7.5 pH, 37°C Optimum Temperature
For Use With (Application) Sequencing
Polymerase Genetically-Engineered form of T7 DNA Polymerase
Product Type Sequenase Version 2.0 DNA Polymerase
Quantity 1000 U
Purity 0.95

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.