Learn More
Hamilton™ Luer Lock Septum Adapter
Click to view available options
:
Includes Septa
Description
- Samples can be added or removed from the sealed container using another syringe to pierce the replaceable septum
- 1 adapter and 12 septa included
- Sampling and storage applications
Specifications
Specifications
| For Use With (Application) | Sampling and storage |
| Includes | 12 replacement septa, 1 adapter |
| Product Type | Septum Adapter |
| Connection Type | Luer |